missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dibutylformamide 98.0+%, TCI America™

Catalog No. D2147500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2147500ML 500 mL
D214725ML 25 mL
D2147100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2147500ML Supplier TCI America Supplier No. D2147500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 761-65-9
Molecular Formula C9H19NO
Molecular Weight (g/mol) 157.26
MDL Number MFCD00003289
InChI Key NZMAJUHVSZBJHL-UHFFFAOYSA-N
Synonym dibutylformamide, formamide, n,n-dibutyl, n,n-di-n-butylformamide, formamide, n,n-di-n-butyl, dibutylamid kyseliny mravenci, dibutylamid kyseliny mravenci czech, formamide, dibutyl, n-formyldibutylamine, formamide,n-dibutyl, n,n-di-butylformamide
PubChem CID 12975
IUPAC Name N,N-dibutylformamide
SMILES CCCCN(CCCC)C=O

Specifications

Color Yellow
Boiling Point 243°C
Quantity 500 mL
Formula Weight 157.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dibutylformamide
RTECSNumber LQ1833000
TSCA Yes