accessibility menu, dialog, popup

UserName

CH3NCH3NNOO(E/Z)

CAS RN 76058-33-8

IUPAC Name: 2-{2-[4-(diethylamino)phenyl]diazen-1-yl}benzoate
Synonyms: 2-{2-[4-(diethylamino)phenyl]diazen-1-yl}benzoate
Molecular Weight (g/mol): 296.35
Molecular Formula: C17H18N3O2
InChi Key: HBRCDTRQDHMTDA-UHFFFAOYSA-M
SMILES: CCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)