missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Red 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 146°C |
| Color | Purple/Red |
| Synonym | 4-(Diethylamino)azobenzene-2′-carboxylic Acid, Diethyl Red |
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 297.36 |
| Percent Purity | ≥97.0% (T) |
| Chemical Name or Material | Ethyl Red [not Cyanin dyes] |
Safety and Handling
| TSCA | No |