missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',7'-Dichlorofluorescein, ACS Reagent, Reagents
Specifications
Specifications
| Boiling Point | 634°C |
| Packaging | Glass Bottle |
| Specific Gravity | 1 |
| pH Color Changes | Greenish-Yellow (excess Chloride) to Rose (excess Silver ions) |
| Color | Orange |
| Quantity | 10 g |
| Physical Form | Solid |
| Grade | ACS |
| Chemical Name or Material | 2',7'-Dichlorofluorescein, ACS Reagent |