accessibility menu, dialog, popup

UserName

CAS RN 76-13-1

FFClFClCl

CAS RN 76-13-1

IUPAC Name: 1,1,2-trichloro-1,2,2-trifluoroethane
Synonyms: 1,1,2-trichlorotrifluoroethane
Molecular Weight (g/mol): 187.37
Molecular Formula: C2Cl3F3
InChi Key: AJDIZQLSFPQPEY-UHFFFAOYSA-N
SMILES: FC(F)(Cl)C(F)(Cl)Cl
Element 
  • (1)
Grade 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)