accessibility menu, dialog, popup

UserName

CAS RN 75695-93-1

IUPAC Name: 3-methyl 5-propan-2-yl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: isradipine isradipine, (R)-isomer (R)-isomer isradipine isradipine (product) dynacire CR (s)-isomer isradipine isradipine, (+-)-isomer isradipine (substance) (+-)-isomer isradipine clivoten
Molecular Weight (g/mol): 371.39
Molecular Formula: C19H21N3O5
InChi Key: HMJIYCCIJYRONP-UHFFFAOYNA-N
SMILES: COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC2=NON=C12)C(=O)OC(C)C
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)