missing translation for 'onlineSavingsMsg'
Learn More

Isradipine 98.0+%, TCI America™

Catalog No. I087625MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I087625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I087625MG Supplier TCI America Supplier No. I087625MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75695-93-1
Molecular Formula C19H21N3O5
Molecular Weight (g/mol) 371.39
MDL Number MFCD00153820
InChI Key HMJIYCCIJYRONP-UHFFFAOYNA-N
Synonym isradipine, dynacirc, isradipin, lomir, isrodipine, prescal, dynacirc cr, esradin, clivoten, dynacrine
PubChem CID 3784
IUPAC Name 3-methyl 5-propan-2-yl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC2=NON=C12)C(=O)OC(C)C

Specifications

Melting Point 169°C
Color Yellow
Quantity 25 mg
Formula Weight 371.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Isradipine
TSCA No
Recommended Storage Freezer (-20 °C)