accessibility menu, dialog, popup

UserName

H3CNNClNHNHNOCH3HCl

CAS RN 75536-04-8

IUPAC Name: 4-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methylpyrimidin-5-amine hydrochloride
Synonyms: moxonidine hydrochloride
Molecular Weight (g/mol): 278.14
Molecular Formula: C9H13Cl2N5O
InChi Key: ZZPAWQYZQVUVHX-UHFFFAOYSA-N
SMILES: Cl.COC1=C(NC2=NCCN2)C(Cl)=NC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results