missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(IV) chloride, 1M soln. in dichloromethane
Description
Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines
Solubility
Reacts with water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines
Solubility
Reacts with water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
Specifications
Specifications
| Quantity | 500 mL |
| Sensitivity | Moisture sensitive; May form a precipitate |
| Density | 1.365 |
| Linear Formula | TiCl4 |
| UN Number | UN2922 |
| Solubility Information | Reacts with water. |
| Formula Weight | 189.69 |
| Concentration or Composition (by Analyte or Components) | 1M soln. in dichloromethane |
| Chemical Name or Material | Titanium(IV) chloride |
Safety and Handling
| Hazard Category | H314-H331-H335-H336-H351 |
| Hazard Statement | GHS H Statement H314-H351 Causes severe skin burns and eye damage. Suspected of causing cancer. |
| Precautionary Statement | P201-P202-P260-P264b-P271-P281-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only