accessibility menu, dialog, popup

UserName

CAS RN 7544-75-4

NNH2HCl

CAS RN 7544-75-4

IUPAC Name: 3,4-dihydro-2H-pyrrol-5-amine hydrochloride
Synonyms: 4,5-dihydro-3H-pyrrol-2-amine hydrochloride
Molecular Weight (g/mol): 120.58
Molecular Formula: C4H9ClN2
InChi Key: JEBHLIYFPKISBI-UHFFFAOYSA-N
SMILES: Cl.NC1=NCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results