missing translation for 'onlineSavingsMsg'
Learn More

Lovastatin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012240
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012240 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012240 Supplier Merck Emd Millipore Supplier No. PHR12851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lovastatin, an orally administered drug, is a lactone metabolite used for the treatment of hypercholesterolemia and coronary heart diseases. It is a hexahydronaphtalene drug, belonging to the class of HMG-CoA (3-hydroxy-3-methyl-glutaryl-coenzyme A) reductase inhibitors.Pharmaceutical secondary standard for application in quality control. Provides pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 75330-75-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C24H36O5
Synonym Lovastatin; Mevinolin
Type Certified Reference Material
Quantity 1 g
Formula Weight 404.54
Grade Certified Reference Material