accessibility menu, dialog, popup

UserName

CAS RN 751-94-0

IUPAC Name: sodium 2-[(1Z,2S,3aS,3bS,5aS,6S,7R,9aS,9bS,10R,11aR)-2-(acetyloxy)-7,10-dihydroxy-3a,3b,6,9a-tetramethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-ylidene]-6-methylhept-5-enoate
Synonyms: sodium 2-[(1Z,2S,3aS,3bS,5aS,6S,7R,9aS,9bS,10R,11aR)-2-(acetyloxy)-7,10-dihydroxy-3a,3b,6,9a-tetramethyl-dodecahydro-2H-cyclopenta[a]phenanthren-1-ylidene]-6-methylhept-5-enoate
Molecular Weight (g/mol): 538.7
Molecular Formula: C31H47NaO6
InChi Key: HJHVQCXHVMGZNC-JCJNLNMISA-M
SMILES: [Na+].C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]1(C)[C@H]2[C@H](O)C[C@H]2\C([C@H](C[C@]12C)OC(C)=O)=C(/CCC=C(C)C)C([O-])=O
Quantity 
  • (1)
  • (1)
Type 
  • (2)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results