missing translation for 'onlineSavingsMsg'
Learn More

Fusidic acid sodium

Catalog No. AAJ6670306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6670306 5 g
AAJ6670303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6670306 Supplier Thermo Scientific Chemicals Supplier No. J6670306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An antitubercular antibiotic which inhibits cell wall formation in Mycobacterium species | CAS: 751-94-0 | C31H47NaO6 | 538.701 g/mol

Targets Gram-positive bacteria namely those which cause skin infections

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 751-94-0
Molecular Formula C31H47NaO6
Molecular Weight (g/mol) 538.701
MDL Number MFCD09054714
InChI Key HJHVQCXHVMGZNC-RDWMJMSASA-M
Synonym Fucidin
PubChem CID 126963539
IUPAC Name sodium;2-[(3R,4S,5S,8S,9R,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoate
SMILES CC1C2CCC3(C(C2(CCC1O)C)C(CC4C3(CC(C4=C(CCC=C(C)C)C(=O)[O-])OC(=O)C)C)O)C.[Na+]

Specifications

Type Fusidic Acid Sodium
Physical Form Powder
Packaging Plastic Bottle
Quantity 5 g
Sensitivity Light and moisture sensitive
Solubility Information Soluble in water
Formula Weight 538.69
Chemical Name or Material Fusidic acid sodium
Hazard Category H302-H315-H319
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 212-030-3
RTECSNumber LV5775000
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only