missing translation for 'onlineSavingsMsg'
Learn More

Chlorotrimethylsilane, 98%

Catalog No. AC110122500
Change view
Click to view available options
Quantity:
25 mL
100 mL
250 mL
1 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110122500 250 mL Glass bottle
AC110120250 25 mL Glass bottle
AC110121000 100 mL Glass bottle
AC110120010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC110122500 Supplier Thermo Scientific Chemicals Supplier No. 110122500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75-77-4 | C3H9ClSi | 108.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent

Chemical Identifiers

CAS 75-77-4
Molecular Formula C3H9ClSi
Molecular Weight (g/mol) 108.64
MDL Number MFCD00000502
InChI Key IJOOHPMOJXWVHK-UHFFFAOYSA-N
Synonym trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmscl
PubChem CID 6397
ChEBI CHEBI:85069
IUPAC Name chloro(trimethyl)silane
SMILES C[Si](C)(C)Cl

Specifications

Melting Point -58.0°C
Color Colorless
Density 0.8500g/mL
Boiling Point 57.0°C
Flash Point −28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3870 to 1.3890
Linear Formula (CH3)3SiCl
Quantity 250 mL
Beilstein 04,IV,4007
Fieser 01,1232; 02,435; 03,310; 04,537; 05,709; 06,626; 07,66; 08,107; 09,112; 10,96; 11,127; 12,279; 13,165; 14,88; 15,14; 16,23; 17,79
Merck Index 15,218
Specific Gravity 0.85
Solubility Information Solubility in water: reacts. Other solubilities: care must be taken with ketones,soluble in benzene,ether and perchloroethylene
Viscosity 0.34 mPa.s (25°C)
Formula Weight 108.64
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chlorotrimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Harmful if inhaled.
Toxic if swallowed.
Harmful in contact with skin.
Reacts violently with water.
Corrosive to the res
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 200-900-5

RUO – Research Use Only