accessibility menu, dialog, popup

UserName

NOCl

CAS RN 74448-92-3

IUPAC Name: 4-(4-chlorophenoxy)benzonitrile
Synonyms: 4-(4-chlorophenoxy)benzonitrile
Molecular Weight (g/mol): 229.66
Molecular Formula: C13H8ClNO
InChi Key: SOTAPBLDXLHNRZ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(OC2=CC=C(C=C2)C#N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results