missing translation for 'onlineSavingsMsg'
Learn More

Terbium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™

Catalog No. CLTB21AY
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLTB21AY 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLTB21AY Supplier SPEX CertiPrep Group LLC Supplier No. CLTB21AY

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Tb10μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Claritas PPT Grade Terbium Standard
Packaging Triple-leached LDPE bottle
Certificate of Analysis Claritas PPT Certificate of Analysis reports up to 69 elements at ppt levels.
CAS 7440-27-9, 7697-37-2, 7732-18-5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Min % 0.0, 2.0, 97.99
CAS Max % 0.0, 2.0, 97.99
Molecular Formula Tb
Technique ICP-MS
MDL Number MFCD00011256
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type ICP/ICP-MS Standard
InChI Key GZCRRIHWUXGPOV-UHFFFAOYSA-N
SMILES [Tb]
Solvent or Matrix 2% HNO3
IUPAC Name terbium
Molecular Weight (g/mol) 158.93
Quantity 125 mL
PubChem CID 23958
ChEBI CHEBI:33376
Grade ICP/ICP-MS
Show More Show Less

Product intended for laboratory use only