accessibility menu, dialog, popup

UserName

CAS RN 7425-80-1

TiH3CCH3OCH3H3COCH3H3COCH3H3CO

CAS RN 7425-80-1

IUPAC Name: titanium(4+) tetrakis(2-methylpropan-1-olate)
Synonyms: titanium(4+) tetrakis(2-methylpropan-1-olate)
Molecular Weight (g/mol): 340.33
Molecular Formula: C16H36O4Ti
InChi Key: QUVMSYUGOKEMPX-UHFFFAOYSA-N
SMILES: [Ti+4].CC(C)C[O-].CC(C)C[O-].CC(C)C[O-].CC(C)C[O-]
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)