Learn More
Titanium(IV) isobutoxide
CAS: 7425-80-1 | C16H36O4Si | 320.545 g/mol
$80.32 - $746.73
Chemical Identifiers
| CAS | 7425-80-1 |
|---|---|
| MDL Number | MFCD00015045 |
| Synonym | tetrakis 2-methylpropyl orthosilicate, titanium iv isobutoxide, tetrakis 2-methylpropyl silicate, tetraisobutyl orthosilicate, silane, tetraisobutoxy, silicic acid tetraisobutyl ester, orthosilicic acid tetrakis 2-methylpropyl ester, silicic acid h4sio4 ,tetrakis 2-methylpropyl ester |
| PubChem CID | 69626 |
| IUPAC Name | tetrakis(2-methylpropyl) silicate |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA8945518
|
Thermo Scientific Chemicals
08945518 |
50 g |
Each for $80.32
|
|
|||||
|
AA8945530
|
Thermo Scientific Chemicals
08945530 |
250 g |
Each for $236.55
|
|
|||||
|
AA89455A1
|
Thermo Scientific Chemicals
089455A1 |
1 kg |
Each for $746.73
|
|
|||||
Description
Titanium(IV) isobutoxide general application in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7425-80-1 | |
| tetrakis 2-methylpropyl orthosilicate, titanium iv isobutoxide, tetrakis 2-methylpropyl silicate, tetraisobutyl orthosilicate, silane, tetraisobutoxy, silicic acid tetraisobutyl ester, orthosilicic acid tetrakis 2-methylpropyl ester, silicic acid h4sio4 ,tetrakis 2-methylpropyl ester | |
| tetrakis(2-methylpropyl) silicate |
| MFCD00015045 | |
| 69626 |
Specifications
| 296°C | |
| Yellow | |
| 50 g | |
| Ti[(CH3)2CHCH2O]4 | |
| 77°C (171°F) | |
| Reacts with water. | |
| CC(C)CO[Si](OCC(C)C)(OCC(C)C)OCC(C)C | |
| 320.545 | |
| 340.34 | |
| Moisture sensitive | |
| Titanium (IV) isobutoxide |
| 7425-80-1 | |
| Melt | |
| C16H36O4Si | |
| MFCD00015045 | |
| tetrakis 2-methylpropyl orthosilicate, titanium iv isobutoxide, tetrakis 2-methylpropyl silicate, tetraisobutyl orthosilicate, silane, tetraisobutoxy, silicic acid tetraisobutyl ester, orthosilicic acid tetrakis 2-methylpropyl ester, silicic acid h4sio4 ,tetrakis 2-methylpropyl ester | |
| QKCMIPQMYBKEPP-UHFFFAOYSA-N | |
| tetrakis(2-methylpropyl) silicate | |
| 69626 | |
| 1.495 | |
| 1.02 g/mL |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 231-061-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only