accessibility menu, dialog, popup

UserName

H3COOCH3CH3I

CAS RN 7425-55-0

IUPAC Name: ethyl 2-iodo-2-methylpropanoate
Synonyms: ethyl 2-iodo-2-methylpropanoate
Molecular Weight (g/mol): 242.06
Molecular Formula: C6H11IO2
InChi Key: AYVTVRNTNCEPSH-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)(C)I
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results