missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Éthyle 2-iodo-2-méthylpropionate 94,0+%, TCI America™
Spécifications
Spécifications
| Quantity | 5 g |
| Formula Weight | 242.06 |
| Percent Purity | ≥94.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Ethyl 2-Iodo-2-methylpropionate |
Sécurité et manipulation
| TSCA | No |
| Recommended Storage | Congélateur |