accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 7424-54-6

IUPAC Name: heptane-3,5-dione
Synonyms: 3,5-heptadione
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: DGCTVLNZTFDPDJ-UHFFFAOYSA-N
SMILES: CCC(=O)CC(=O)CC
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)