missing translation for 'onlineSavingsMsg'
Learn More

3,5-Heptanedione, 97%

Catalog No. AAA1816514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1816514 25 g
AAA1816506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1816514 Supplier Thermo Scientific Chemicals Supplier No. A1816514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7424-54-6 | C7H12O2 | 128.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7424-54-6
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00015186
InChI Key DGCTVLNZTFDPDJ-UHFFFAOYSA-N
Synonym 3,5-heptanedione, dipropionylmethane, 3,5-heptandione, unii-q3b558e3vy, 3,5-heptadione, 3,5-heptane dione, pubchem21961, ksc379q6f, 3,5-heptanedione, purum gc
PubChem CID 81923
IUPAC Name heptane-3,5-dione
SMILES CCC(=O)CC(=O)CC

Specifications

Density 0.8
Boiling Point 175°C to 177°C
Flash Point 57°C (134°F)
Odor Pleasant
Refractive Index 1.4555
Quantity 25 g
UN Number UN1224
Beilstein 635979
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material 3,5-Heptanedione
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 231-054-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only