missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-L-tyrosine 98.0+%, TCI America™

Catalog No. F0201100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0201100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0201100MG Supplier TCI America Supplier No. F0201100MG

Chemical Identifiers

CAS 7423-96-3
Molecular Formula C9H10FNO3
Molecular Weight (g/mol) 199.18
MDL Number MFCD00002607
InChI Key VIIAUOZUUGXERI-JLDDOWRYNA-N
Synonym 3-fluoro-l-tyrosine, s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluorotyrosine, 3-fluorotyrosine, l, h-tyr 3-f-oh, unii-174nrg3m2x, s-3-fluorotyrosine, 3'-fluoro-l-tyrosine, 2s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluoro-tyrosine
PubChem CID 643330
ChEBI CHEBI:46534
IUPAC Name (2S)-2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(F)=C1)C(O)=O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 199.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-L-tyrosine