accessibility menu, dialog, popup

UserName

CAS RN 7413-34-5

IUPAC Name: disodium 2-[(4-{[(2,4-diaminopteridin-6-yl)methyl](methyl)amino}phenyl)formamido]pentanedioate
Synonyms: disodium 2-[(4-{[(2,4-diaminopteridin-6-yl)methyl](methyl)amino}phenyl)formamido]pentanedioate
Molecular Weight (g/mol): 498.41
Molecular Formula: C20H20N8Na2O5
InChi Key: DASQOOZCTWOQPA-UHFFFAOYNA-L
SMILES: [Na+].[Na+].CN(CC1=CN=C2N=C(N)N=C(N)C2=N1)C1=CC=C(C=C1)C(=O)NC(CCC([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results