accessibility menu, dialog, popup

UserName

CAS RN 74115-13-2

OHNBr

CAS RN 74115-13-2

IUPAC Name: 5-bromopyridin-3-ol
Synonyms: 5-bromopyridin-3-ol
Molecular Weight (g/mol): 174
Molecular Formula: C5H4BrNO
InChi Key: VNYBIBSZZDAEOK-UHFFFAOYSA-N
SMILES: OC1=CN=CC(Br)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)