missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-hydroxypyridine 98.0+%, TCI America™
$61.51 - $201.32
Chemical Identifiers
| CAS | 74115-13-2 |
|---|---|
| Molecular Formula | C5H4BrNO |
| Molecular Weight (g/mol) | 174.00 |
| MDL Number | MFCD00661305 |
| InChI Key | VNYBIBSZZDAEOK-UHFFFAOYSA-N |
| Synonym | 3-bromo-5-hydroxypyridine, 5-bromo-3-pyridinol, 5-bromo-3-hydroxypyridine, 5-bromo-pyridin-3-ol, 3-pyridinol, 5-bromo, 3-bromo-5-pyridinol, 3-hydroxy-5-bromopyridine, 5-bromo-3-hydroxy-pyridine, pyridine, 3-ol-5-bromo, 5-bromopyrid-3-ol |
| PubChem CID | 599529 |
| IUPAC Name | 5-bromopyridin-3-ol |
| SMILES | OC1=CN=CC(Br)=C1 |
Chemical Identifiers
| 74115-13-2 | |
| 174.00 | |
| VNYBIBSZZDAEOK-UHFFFAOYSA-N | |
| 599529 | |
| OC1=CN=CC(Br)=C1 |
| C5H4BrNO | |
| MFCD00661305 | |
| 3-bromo-5-hydroxypyridine, 5-bromo-3-pyridinol, 5-bromo-3-hydroxypyridine, 5-bromo-pyridin-3-ol, 3-pyridinol, 5-bromo, 3-bromo-5-pyridinol, 3-hydroxy-5-bromopyridine, 5-bromo-3-hydroxy-pyridine, pyridine, 3-ol-5-bromo, 5-bromopyrid-3-ol | |
| 5-bromopyridin-3-ol |
Specifications
| 74115-13-2 | |
| White-Yellow | |
| MFCD00661305 | |
| 3-bromo-5-hydroxypyridine, 5-bromo-3-pyridinol, 5-bromo-3-hydroxypyridine, 5-bromo-pyridin-3-ol, 3-pyridinol, 5-bromo, 3-bromo-5-pyridinol, 3-hydroxy-5-bromopyridine, 5-bromo-3-hydroxy-pyridine, pyridine, 3-ol-5-bromo, 5-bromopyrid-3-ol | |
| OC1=CN=CC(Br)=C1 | |
| 174.00 | |
| 174.00 | |
| Crystalline Powder |
| 169°C | |
| C5H4BrNO | |
| 1 g | |
| VNYBIBSZZDAEOK-UHFFFAOYSA-N | |
| 5-bromopyridin-3-ol | |
| 599529 | |
| ≥98.0% (GC,T) | |
| 3-Bromo-5-hydroxypyridine |
Safety and Handling
TSCA : No