accessibility menu, dialog, popup

UserName

CAS RN 741-58-2

CH3H3COPSOCH3H3CSNHSOO

CAS RN 741-58-2

IUPAC Name: O,O-bis(propan-2-yl) [(2-benzenesulfonamidoethyl)sulfanyl]phosphonothioate
Synonyms: bensulide
Molecular Weight (g/mol): 397.5
Molecular Formula: C14H24NO4PS3
InChi Key: RRNIZKPFKNDSRS-UHFFFAOYSA-N
SMILES: CC(C)OP(=S)(OC(C)C)SCCNS(=O)(=O)C1=CC=CC=C1
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results