missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Bensulide Analytical Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 741-58-2 |
| Physical Form | Neat |
| Molecular Formula | C14H24NO4PS3 |
| Linear Formula | C14H24NO4PS3 |
| MDL Number | MFCD00055362 |
| UN Number | UN 3077 9 / PGIII |
| Quantity | 100 mg |
| Formula Weight | 397.51 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH