accessibility menu, dialog, popup

UserName

OBrBr

CAS RN 74003-55-7

IUPAC Name: 3,4-dibromobenzaldehyde
Synonyms: 3,4-dibromobenzaldehyde
Molecular Weight (g/mol): 263.92
Molecular Formula: C7H4Br2O
InChi Key: UTYRZXNEFUYFCJ-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results