accessibility menu, dialog, popup

UserName

H3CNCH3NOO

CAS RN 7385-67-3

IUPAC Name: 8-(diethylamino)-12H-10-oxa-5-azatetraphen-12-one
Synonyms: nile red nile red dye
Molecular Weight (g/mol): 318.38
Molecular Formula: C20H18N2O2
InChi Key: VOFUROIFQGPCGE-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31
Color 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)