accessibility menu, dialog, popup

UserName

CH3OOClO

CAS RN 7377-26-6

IUPAC Name: methyl 4-(carbonochloridoyl)benzoate
Synonyms: methyl 4-(carbonochloridoyl)benzoate
Molecular Weight (g/mol): 198.6
Molecular Formula: C9H7ClO3
InChi Key: CVXXHXPNTZBZEL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C(Cl)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)