missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(Chloroformyl)benzoate 95.0+%, TCI America™

Catalog No. T0283500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0283500G 500 g
T028325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0283500G Supplier TCI America Supplier No. T0283500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7377-26-6
Molecular Formula C9H7ClO3
Molecular Weight (g/mol) 198.602
MDL Number MFCD00059486
InChI Key CVXXHXPNTZBZEL-UHFFFAOYSA-N
Synonym methyl 4-chlorocarbonyl benzoate, methyl 4-chlorocarbonylbenzoate, methyl 4-chloroformyl benzoate, terephthalic acid monomethyl ester chloride, methyl 4-carbonochloridoyl benzoate, methyl terephthaloyl chloride, benzoic acid, 4-chlorocarbonyl-, methyl ester, 4-chlorocarbonyl-benzoic acid methyl ester, 4-carbomethoxybenzoylchloride, 4-chlorocarbonylbenzoic acid methyl ester
PubChem CID 81850
IUPAC Name methyl 4-carbonochloridoylbenzoate
SMILES COC(=O)C1=CC=C(C=C1)C(=O)Cl

Specifications

Melting Point 54°C
Boiling Point 140°C
Quantity 500 g
UN Number 3261
Formula Weight 198.60
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(Chloroformyl)benzoate
EINECSNumber (3)-3502
TSCA No