missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chlorocarbonylbenzoate, 90%, tech.

Catalog No. AC424710010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424710010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC424710010 Supplier Thermo Scientific Chemicals Supplier No. 424710010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7377-26-6 | C9H7ClO3 | 198.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7377-26-6
Molecular Formula C9H7ClO3
Molecular Weight (g/mol) 198.61
MDL Number MFCD00059486
InChI Key CVXXHXPNTZBZEL-UHFFFAOYSA-N
Synonym methyl 4-chlorocarbonyl benzoate, methyl 4-chlorocarbonylbenzoate, methyl 4-chloroformyl benzoate, terephthalic acid monomethyl ester chloride, methyl 4-carbonochloridoyl benzoate, methyl terephthaloyl chloride, benzoic acid, 4-chlorocarbonyl-, methyl ester, 4-chlorocarbonyl-benzoic acid methyl ester, 4-carbomethoxybenzoylchloride, 4-chlorocarbonylbenzoic acid methyl ester
PubChem CID 81850
IUPAC Name methyl 4-carbonochloridoylbenzoate
SMILES COC(=O)C1=CC=C(C=C1)C(=O)Cl

Specifications

Melting Point 50°C to 59°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula H3COOCC6H4COCl
Quantity 1 g
Solubility Information Solubility in water: hydrolysis
Formula Weight 198.61
Percent Purity 90%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-chlorocarbonylbenzoate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 230-937-2

RUO – Research Use Only