accessibility menu, dialog, popup

UserName

CAS RN 73346-79-9

LiH3COOHOOHOOOOCH3H3COH(E/Z)

CAS RN 73346-79-9

IUPAC Name: lithium(1+) 9-{[4-(3,4-dihydroxy-5-{[3-(3-hydroxybutan-2-yl)oxiran-2-yl]methyl}oxan-2-yl)-3-methylbut-2-enoyl]oxy}nonanoate
Synonyms: lithium(1+) 9-{[4-(3,4-dihydroxy-5-{[3-(3-hydroxybutan-2-yl)oxiran-2-yl]methyl}oxan-2-yl)-3-methylbut-2-enoyl]oxy}nonanoate
Molecular Weight (g/mol): 506.56
Molecular Formula: C26H43LiO9
InChi Key: XFIKMPRBSORKQP-UHFFFAOYNA-M
SMILES: [Li+].CC(O)C(C)C1OC1CC1COC(CC(C)=CC(=O)OCCCCCCCCC([O-])=O)C(O)C1O
Quantity 
  • (1)
Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results