missing translation for 'onlineSavingsMsg'
Learn More

Mupirocin lithium salt, 90+%

Catalog No. AAJ66890MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66890MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66890MC Supplier Thermo Scientific Chemicals Supplier No. J66890MC

Commonly used to target gram-positive bacteria | CAS: 73346-79-9 | C26H43LiO9 | 506.561 g/mol

Commonly used to target Gram-positive bacteria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Type Mupirocin Lithium Salt
Physical Form Powder
CAS 73346-79-9
Packaging Plastic Bottle
Quantity 100 mg
Molecular Formula C26H43LiO9
MDL Number MFCD11977847
Solubility Information Soluble in water at approximately 10mg/ml
InChI Key XFIKMPRBSORKQP-JATHGWPISA-M
SMILES [Li+].CC(C1C(O1)CC2COC(C(C2O)O)CC(=CC(=O)OCCCCCCCCC(=O)[O-])C)C(C)O
IUPAC Name lithium;9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoate
Molecular Weight (g/mol) 506.561
PubChem CID 71311795
Formula Weight 506.55
Percent Purity 95%
Chemical Name or Material Mupirocin lithium salt
Show More Show Less
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only