missing translation for 'onlineSavingsMsg'
Learn More

Dibromomalonamide, 98+%

Catalog No. AAB2231522
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2231522 100 g
AAB2231509 10 g
AAB2231514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2231522 Supplier Thermo Scientific Chemicals Supplier No. B2231522

CAS: 73003-80-2 | C3H4Br2N2O2 | 259.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Dibromomalonamide is an antimicrobial agent used in sugar mills, a compound for lipophilic fluid systems for fabric care. It is also used to produce 5-thioxo-[1,3]dithiolo[4,5-d][1,3]dithiole-2,2-dicarboxylic acid diamide.

Solubility
Soluble in water.

Notes
Moisture & light sensitive. Store away from oxidizing agent.

Chemical Identifiers

CAS 73003-80-2
Molecular Formula C3H4Br2N2O2
Molecular Weight (g/mol) 259.89
MDL Number MFCD00014803
InChI Key SWHQVMGRXIYDSF-UHFFFAOYSA-N
PubChem CID 175375
SMILES NC(=O)C(Br)(Br)C(N)=O

Specifications

Melting Point 202°C to 207°C
Quantity 100 g
Beilstein 1770873
Sensitivity Light and moisture sensitive
Synonym dibromomalonamide, 2,2-dibromomalonamide, dibromopropanediamide, propanediamide, 2,2-dibromo, dibromo malonamide-13c3, dibromo malonamide, dibromomalonic diamide, dibromomalonic acid diamide, propanediamide,2,2-dibromo, 2,2-bis bromanyl propanediamide
Solubility Information Soluble in water.
IUPAC Name 2,2-dibromopropanediamide
Formula Weight 259.89
Percent Purity ≥98%
Chemical Name or Material Dibromomalonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 277-205-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only