missing translation for 'onlineSavingsMsg'
Learn More

Dibromomalonamide 98.0+%, TCI America™

Catalog No. D42725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42725G 5 g
D427225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42725G Supplier TCI America Supplier No. D42725G

Chemical Identifiers

CAS 73003-80-2
Molecular Formula C3H4Br2N2O2
Molecular Weight (g/mol) 259.89
MDL Number MFCD00014803
InChI Key SWHQVMGRXIYDSF-UHFFFAOYSA-N
Synonym dibromomalonamide, 2,2-dibromomalonamide, dibromopropanediamide, propanediamide, 2,2-dibromo, dibromo malonamide-13c3, dibromo malonamide, dibromomalonic diamide, dibromomalonic acid diamide, propanediamide,2,2-dibromo, 2,2-bis bromanyl propanediamide
PubChem CID 175375
IUPAC Name dibromopropanediamide
SMILES NC(=O)C(Br)(Br)C(N)=O

Specifications

Melting Point 204°C
Color White
Quantity 5 g
Formula Weight 259.89
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Dibromomalonamide