CAS RN 730-40-5
CAS RN 730-40-5
IUPAC Name:
4-[2-(4-nitrophenyl)diazen-1-yl]aniline
Synonyms:
4-[2-(4-nitrophenyl)diazen-1-yl]aniline
Molecular Weight (g/mol):
242.24
Molecular Formula:
C12H10N4O2
InChi Key:
UNBOSJFEZZJZLR-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1)N=NC1=CC=C(C=C1)[N+]([O-])=O
1
–
2
of
2
results
1
Disperse Orange 3, ≥96% (HPLC), MilliporeSigma™ Supelco™
Disperse Orange 3 is widely used in textile industry for dyeing polyester fibers.
2
Thermo Scientific Chemicals Disperse Orange 3, 65%
CAS: 730-40-5 Molecular Formula: C12H10N4O2 Molecular Weight (g/mol): 242.238 MDL Number: MFCD00007856 InChI Key: UNBOSJFEZZJZLR-UHFFFAOYSA-N Synonym: C.I. 11005 PubChem CID: 12896 ChEBI: CHEBI:75648 IUPAC Name: 4-[(4-nitrophenyl)diazenyl]aniline SMILES: C1=CC(=CC=C1N)N=NC2=CC=C(C=C2)[N+](=O)[O-]