missing translation for 'onlineSavingsMsg'
Learn More

Disperse Orange 3, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015367
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015367 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015367 Supplier Merck Emd Millipore Supplier No. 5388225MG

missing translation for 'validMainframeMsg'

Disperse Orange 3 is widely used in textile industry for dyeing polyester fibers.

Used to provide experimental verification of a two-level model for the enhancement of the nonresonant, nonlinear refractive index, with a decrease in absorption by reduction of a donor group strength. Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 730-40-5
Physical Form Neat
Linear Formula O2 NC6H4 N=NC6H4 NH2
MDL Number MFCD00007856
Melting Point 200°C
Synonym 4-(4-Nitrophenylazo)aniline
Type Analytical Standard
Quantity 25 mg
Formula Weight 242.23
Percent Purity ≥96% (HPLC)
Grade Analytical Standard
Show More Show Less