accessibility menu, dialog, popup

UserName

H3CNNSCH3

CAS RN 72987-62-3

IUPAC Name: 2-ethyl-3-(methylsulfanyl)pyrazine
Synonyms: 2-ethyl-3-(methylsulfanyl)pyrazine
Molecular Weight (g/mol): 154.23
Molecular Formula: C7H10N2S
InChi Key: XYHPPOMSLGJAAM-UHFFFAOYSA-N
SMILES: CCC1=NC=CN=C1SC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results