accessibility menu, dialog, popup

UserName

FFFFFFFFF

CAS RN 729-81-7

IUPAC Name: 1,3,5-tris(trifluoromethyl)benzene
Synonyms: 1,3,5-tris(trifluoromethyl)benzene
Molecular Weight (g/mol): 282.11
Molecular Formula: C9H3F9
InChi Key: ZMAUHKSOLPYPDB-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results