accessibility menu, dialog, popup

UserName

CAS RN 7269-35-4

IUPAC Name: (butylsulfanyl)(phenyl)methanone
Synonyms: (butylsulfanyl)(phenyl)methanone
Molecular Weight (g/mol): 194.29
Molecular Formula: C11H14OS
InChi Key: KXXZPIFZTPWOJJ-UHFFFAOYSA-N
SMILES: CCCCSC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results