accessibility menu, dialog, popup

UserName

CAS RN 7212-44-4

IUPAC Name: (6E)-3,7,11-trimethyldodeca-1,6,10-trien-3-ol
Synonyms: nerolidol isomers
Molecular Weight (g/mol): 222.37
Molecular Formula: C15H26O
InChi Key: FQTLCLSUCSAZDY-SDNWHVSQNA-N
SMILES: CC(C)=CCC\C(C)=C\CCC(C)(O)C=C
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)