missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene, 97+% (total alcohols), Thermo Scientific™

Catalog No. p-3758208
Change view
Click to view available options
Quantity:
25g
100g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC121970250 25g
AC121971000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC121970250 Supplier Thermo Scientific Chemicals Supplier No. 121970250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7212-44-4
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.37
MDL Number MFCD00008911,MFCD00008911,MFCD00008911,MFCD00085350
InChI Key FQTLCLSUCSAZDY-SDNWHVSQNA-N
Synonym 3r,6e-nerolidol, unii-uoc0644v25, 3r-6e-nerolidol, nerolidol, 3r,6e-3,7,11-trimethyldodeca-1,6,10-trien-3-ol, e-nerolidol, 1,6,10-dodecatrien-3-ol, 3,7,11-trimethyl-, 3r,6e, nerolidol, 6e--, --nerolidol, ?-nerolidol
PubChem CID 11241545
ChEBI CHEBI:59959
IUPAC Name (3R,6E)-3,7,11-trimethyldodeca-1,6,10-trien-3-ol
SMILES CC(C)=CCC\C(C)=C\CCC(C)(O)C=C

Specifications

Density 0.8700g/mL
Boiling Point 114.0°C (1.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4780 to 1.4830
Quantity 25g
Specific Gravity 0.87
Merck Index 15, 6561
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in abs. alc. and fixed oils, (still clearly sol. in 3 parts of 70% alc.), insoluble in glycerol
Formula Weight 222.37
Percent Purity 97+% (total alcohols)
Chemical Name or Material 3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene, ≥97% (total alcohols)
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 230-597-5

RUO – Research Use Only