missing translation for 'onlineSavingsMsg'
Learn More

Nerolidol (cis- and trans- mixture) 97.0+%, TCI America™

Catalog No. N045425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N045425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N045425ML Supplier TCI America Supplier No. N045425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7212-44-4
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.37
MDL Number MFCD00008911,MFCD00008911,MFCD00008911,MFCD00085350
InChI Key FQTLCLSUCSAZDY-SDNWHVSQNA-N
Synonym 3r,6e-nerolidol, unii-uoc0644v25, 3r-6e-nerolidol, nerolidol, 3r,6e-3,7,11-trimethyldodeca-1,6,10-trien-3-ol, e-nerolidol, 1,6,10-dodecatrien-3-ol, 3,7,11-trimethyl-, 3r,6e, nerolidol, 6e--, --nerolidol, ?-nerolidol
PubChem CID 11241545
ChEBI CHEBI:59959
IUPAC Name (6E)-3,7,11-trimethyldodeca-1,6,10-trien-3-ol
SMILES CC(C)=CCC\C(C)=C\CCC(C)(O)C=C

Specifications

Color Yellow
Boiling Point 146°C
Quantity 25 mL
Formula Weight 222.37
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Nerolidol (cis- and trans- mixture)
RTECSNumber JR4977000
TSCA Yes