accessibility menu, dialog, popup

UserName

CAS RN 7170-38-9

IUPAC Name: 3-phenoxypropanoate
Synonyms: β-phenoxypropionate
Molecular Weight (g/mol): 165.17
Molecular Formula: C9H9O3
InChi Key: BUSOTUQRURCMCM-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCOC1=CC=CC=C1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)