accessibility menu, dialog, popup

UserName

CAS RN 71242-58-5

H3COHHSH3C

CAS RN 71242-58-5

IUPAC Name: 7,7-dimethyl-1-(sulfanylmethyl)bicyclo[2.2.1]heptan-2-ol
Synonyms: 7,7-dimethyl-1-(sulfanylmethyl)bicyclo[2.2.1]heptan-2-ol
Molecular Weight (g/mol): 186.31
Molecular Formula: C10H18OS
InChi Key: PYQMNINTTPIRIT-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(CS)C(O)C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results