missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S)-(-)-10-Mercaptoisoborneol 98.0+%, TCI America™
Supplier: TCI America M10701G
Specifications
| (1S)-(-)-10-Mercaptoisoborneol | |
| 72°C | |
| C10H18OS | |
| 1 g | |
| CC1(C2CCC1(C(C2)O)CS)C | |
| 186.313 | |
| 186.31 | |
| Crystalline Powder |
| 71242-58-5 | |
| White | |
| MFCD00151646 | |
| PYQMNINTTPIRIT-CCNFQMFXSA-N | |
| (3R)-7,7-dimethyl-4-(sulfanylmethyl)bicyclo[2.2.1]heptan-3-ol | |
| 13460477 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 71242-58-5 | |
| 186.313 | |
| PYQMNINTTPIRIT-CCNFQMFXSA-N | |
| (3R)-7,7-dimethyl-4-(sulfanylmethyl)bicyclo[2.2.1]heptan-3-ol |
| C10H18OS | |
| MFCD00151646 | |
| 13460477 | |
| CC1(C2CCC1(C(C2)O)CS)C |
Safety and Handling
TSCA : No