missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015321
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015321 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015321 Supplier Merck Emd Millipore Supplier No. PHR11081G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. L-Histidine is an essential amino acid, widely present at the active sites of enzymes. It binds to metal ions and may aid in the transport of copper. It also plays an important role for the binding of heme groups, macromolecules and phosphate groups in the binding sites of proteins.

Specifications

CAS 71-00-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C6H9N3O2
MDL Number MFCD00064315
Melting Point 282°C
Synonym (S)-2-Amino-3-(4-imidazolyl)propionic acid; NSC 137773
Type Certified Reference Material
Quantity 1 g
Formula Weight 155.15
Grade Certified Reference Material
Show More Show Less