accessibility menu, dialog, popup

UserName

CAS RN 70831-56-0

OHO(R)OOHOOH(R)OOOHOHOHO(E)(E)

CAS RN 70831-56-0

IUPAC Name: (2R,3R)-2,3-bis({[(2E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy})butanedioic acid
Synonyms: cichoric acid
Molecular Weight (g/mol): 474.37
Molecular Formula: C22H18O12
InChi Key: YDDGKXBLOXEEMN-IABMMNSOSA-N
SMILES: OC(=O)[C@H](OC(=O)\C=C\C1=CC=C(O)C(O)=C1)[C@@H](OC(=O)\C=C\C1=CC=C(O)C(O)=C1)C(O)=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results